Få mere at vide
6-Chloro-7-methylchromone, 98%, Thermo Scientific™
Artikelnummer. 11451640
Skift visning
Mængde:
1 g
5 g
Pakningsstørrelse:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
11451640 5 g 5g
11441640 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11451640 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H26997.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 67029-84-9
Molekylær formel C10H7ClO2
Molekylvægt (g/mol) 194.614
MDL nummer MFCD00239401
InChI nøgle UQXYHMICNLSDMN-UHFFFAOYSA-N
Synonym 6-chloro-7-methylchromone, 6-chloro-7-methyl-4h-chromen-4-one, 6-chloro-7-methyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one,6-chloro-7-methyl
PubChem CID 688707
IUPAC navn 6-chloro-7-methylchromen-4-one
SMIL CC1=C(C=C2C(=O)C=COC2=C1)Cl
Tekniske data
Kemisk navn eller materiale 6-Chloro-7-methylchromone
Smeltepunkt 169°C to 172°C
Mængde 5 g
Formel vægt 194.62
Procent renhed 98%