Få mere at vide
6-Chloro-6-defluoro Ciprofloxacin, TRC
Artikelnummer. 30135572
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135572 1 mg
30011638 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135572 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3680351MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 93106-58-2
Molekylær formel C17H18ClN3O3
Molekylvægt (g/mol) 347.8
Synonym 6-Chloro-1-cyclopropyl-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic Acid
IUPAC navn 6-chloro-1-cyclopropyl-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid
SMIL OC(=O)C1=CN(C2CC2)c3cc(N4CCNCC4)c(Cl)cc3C1=O
Tekniske data
Kemisk navn eller materiale 6-Chloro-6-defluoro Ciprofloxacin
Mængde 1 mg
InChI formel InChI=1S/C17H18ClN3O3/c18-13-7-11-14(8-15(13)20-5-3-19-4-6-20)21(10-1-2-10)9-12(16(11)22)17(23)24/h7-10,19H,1-6H2,(H,23,24)
Formel vægt 347.1037