Få mere at vide
6-Chloro-5-(2-chloroacetyl)indolin-2-one, Mikromol™
Artikelnummer. 30459342
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30459342 4 x 25 mg
30051384 25 mg
30049566 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30459342 Leverandør Mikromol Leverandørnr. MM1273.0800254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6-Chloro-5-(2-chloroacetyl)indolin-2-one, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Chloroacetyl-6-chloro-1,3-dihydroindol-2H-indone-2-one
CAS 118307-04-3
Analyt- eller komponentnavne 5-Chloroacetyl-6-chloro-1,3-dihydroindol-2H-indone-2-one
Forsendelsestilstand Room Temperature
Molekylær formel C10 H7 Cl2 N O2
Certificeringer/compliance ISO 9001
Synonym 6-Chloro-5-(2-chloroacetyl)-1,3-dihydro-2H-indol-2-one, 2H-Indol-2-one, 6-chloro-5-(chloroacetyl)-1,3-dihydro- (9CI), 5-(2-Chloroacetyl)-6-chloro-2-oxindole, 5-(2-Chloroacetyl)-6-chlorooxindole, 6-Chloro-5-(2-chloroacetyl)-1,3-dihydro-2H-indol-2-one, 6-Chloro-5-(2-chloroacetyl)indolin-2-one, 6-Chloro-5-(chloroacetyl)oxindole, 6-Chloro-5-chloroacetyl-2-oxoindoline
InChI formel InChI=1S/C10H7Cl2NO2/c11-4-9(14)6-1-5-2-10(15)13-8(5)3-7(6)12/h1,3H,2,4H2,(H,13,15)
Løsningstype Neat
SMIL ClCC(=O)c1cc2CC(=O)Nc2cc1Cl
IUPAC navn 6-chloro-5-(2-chloroacetyl)-1,3-dihydroindol-2-one
Molekylvægt (g/mol) 244.07
Mængde 4 x 25 mg
Formel vægt 242.9854 g/mol