Få mere at vide
6-Chloro-5-(2-chloro-1-hydroxyethyl)-1,3-dihydro-2H-indol-2-one, TRC
Artikelnummer. 30007759
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30007759 5 mg
30009833 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30007759 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3633735MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 884305-06-0
Molekylær formel C10 H9 Cl2 N O2
Molekylvægt (g/mol) 246.09
Synonym 2H-Indol-2-one, 6-chloro-5-(2-chloro-1-hydroxyethyl)-1,3-dihydro-, 6-Chloro-5-(2-chloro-1-hydroxyethyl)-1,3-dihydro-2H-indol-2-one, 6-Chloro-5-(2-chloro-1-hydroxyethyl)indolin-2-one
IUPAC navn 6-chloro-5-(2-chloro-1-hydroxyethyl)-1,3-dihydroindol-2-one
SMIL O=C1NC2=C(C=C(C(O)CCl)C(Cl)=C2)C1
Tekniske data
Kemisk navn eller materiale 6-Chloro-5-(2-chloro-1-hydroxyethyl)indolin-2-one
Mængde 5 mg
InChI formel InChI=1S/C10H9Cl2NO2/c11-4-9(14)6-1-5-2-10(15)13-8(5)3-7(6)12/h1,3,9,14H,2,4H2,(H,13,15)
Formel vægt 245.001