Learn More
6-Chloro-4-hydroxy-2-methyl-N-(pyridin-2-yl)-2H-thieno[2,3-e][1,2]thiazine-3-carboxamide 1,1-dioxide, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 70374-39-9 |
| Molecular Formula | C13H10ClN3O4S2 |
| MDL Number | MFCD00866163 |
| Quantity | 250 mg |
| SMILES | O=C(C1=C(O)C2=C(C=C(Cl)S2)S(N1C)(=O)=O)NC3=NC=CC=C3 |
| Molecular Weight (g/mol) | 371.82 |
| Percent Purity | 98% |
| Chemical Name or Material | 6-Chloro-4-hydroxy-2-methyl-N-(pyridin-2-yl)-2H-thieno[2,3-e][1,2]thiazine-3-carboxamide 1,1-dioxide |