missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-4-(furfurylamino)pyrimidine, 96%, Thermo Scientific™

Catalog Number 15467177
Click to view available options
Quantity:
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 99846-86-3
Melting Point 128°C to 132°C
Molecular Formula C9H8ClN3O
MDL Number MFCD11655644
Quantity 500 mg
Synonym 6-chloro-n-furan-2-ylmethyl pyrimidin-4-amine, n-6-chloro-4-pyrimidinyl-n-2-furylmethyl amine, 6-chloro-4-furfurylamino pyrimidine
InChI Key STRGJGAQEVTISA-UHFFFAOYSA-N
SMILES C1=COC(=C1)CNC2=CC(=NC=N2)Cl
IUPAC Name 6-chloro-N-(furan-2-ylmethyl)pyrimidin-4-amine
Molecular Weight (g/mol) 209.633
PubChem CID 305494
Formula Weight 209.64
Percent Purity 96%
Chemical Name or Material 6-Chloro-4-(furfurylamino)pyrimidine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'