Få mere at vide
6-Chloro-4-(2-fluorophenyl)-2-methylquinazoline, Mikromol™
Artikelnummer. 30050675
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050675 25 mg
30052019 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050675 Leverandør Mikromol Leverandørnr. MM1106.080025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6-Chloro-4-(2-fluorophenyl)-2-methylquinazoline
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Chloro-4-(2-fluorophenyl)-2-methylquinazoline
CAS 119401-13-7
Analyt- eller komponentnavne 6-Chloro-4-(2-fluorophenyl)-2-methylquinazoline
Forsendelsestilstand Room Temperature
Molekylær formel C15 H10 Cl F N2
Certificeringer/compliance ISO 17025
Synonym 6-Chloro-4-(2-fluorophenyl)-2-methylquinazoline, Midazolam Imp. H (EP), Quinazoline, 6-chloro-4-(2-fluorophenyl)-2-methyl-, 6-Chloro-2-methyl-4-(2-fluorophenyl)quinazoline, Midazolam Impurity H
InChI formel InChI=1S/C15H10ClFN2/c1-9-18-14-7-6-10(16)8-12(14)15(19-9)11-4-2-3-5-13(11)17/h2-8H,1H3
SMIL Cc1nc(c2ccccc2F)c3cc(Cl)ccc3n1
IUPAC navn 6-chloro-4-(2-fluorophenyl)-2-methylquinazoline
Molekylvægt (g/mol) 272.7
Mængde 25 mg
Formel vægt 272.0517 g/mol