Få mere at vide
6-Chloro-4-(2-chlorophenyl)-2-quinazolinecarboxylic Acid, LoGiCal
Artikelnummer. 30462982
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30462982 10 mg
1 options
Artikelnummer. 30462982 Leverandør LoGiCal Leverandørnr. LGCFOR0071.04
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Chloro-4-(2-chlorophenyl)-2-quinazolinecarboxylic Acid
CAS 54643-79-7
Analyt- eller komponentnavne 6-Chloro-4-(2-chlorophenyl)-2-quinazolinecarboxylic Acid
Forsendelsestilstand Room Temperature
Molekylær formel C15 H8 Cl2 N2 O2
Certificeringer/compliance ISO 9001
Synonym 6-Chloro-4-(2-chlorophenyl)-2-quinazolinecarboxylic Acid, Lorazepam Related Compound D
InChI formel InChI=1S/C15H8Cl2N2O2/c16-8-5-6-12-10(7-8)13(19-14(18-12)15(20)21)9-3-1-2-4-11(9)17/h1-7H,(H,20,21)
Løsningstype Neat
SMIL OC(=O)c1nc(c2ccccc2Cl)c3cc(Cl)ccc3n1
IUPAC navn 6-chloro-4-(2-chlorophenyl)quinazoline-2-carboxylic acid
Industry Type Clinical Testing
Molekylvægt (g/mol) 319.14
Mængde 10 mg
Formel vægt 317.9963 g/mol