Få mere at vide
6-Chloro-4-(2-chlorophenyl)-2(1H)-quinazolinone, Mikromol™
Artikelnummer. 17902176
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17902176 100 mg
17962506 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17902176 Leverandør Mikromol Leverandørnr. MM0071.09
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6-Chloro-4-(2-chlorophenyl)-2(1H)-quinazolinone
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Chloro-4-(2-chlorophenyl)-2(1H)-quinazolinone
CAS 23441-87-4
Analyt- eller komponentnavne 6-Chloro-4-(2-chlorophenyl)-2(1H)-quinazolinone
Forsendelsestilstand Room Temperature
Molekylær formel C14 H8 Cl2 N2 O
Certificeringer/compliance ISO 17025
Synonym 6-Chloro-4-(2-chlorophenyl)-2(1H)-quinazolinone
InChI formel InChI=1S/C14H8Cl2N2O/c15-8-5-6-12-10(7-8)13(18-14(19)17-12)9-3-1-2-4-11(9)16/h1-7H,(H,17,18,19)
SMIL Clc1ccc2NC(=O)N=C(c3ccccc3Cl)c2c1
IUPAC navn 6-chloro-4-(2-chlorophenyl)-1H-quinazolin-2-one
Molekylvægt (g/mol) 291.13
Mængde 100 mg
Formel vægt 290.0014 g/mol