Få mere at vide
6-Chloro-3,4-dihydro-4-methyl-3-oxo-2H-1,4-benzoxazine-8-carboxylic Acid-13C,d3 Methyl Ester, TRC
Artikelnummer. 30354110
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30354110 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30354110 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36530310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Chloro-3,4-dihydro-4-methyl-3-oxo-2H-1,4-benzoxazine-8-carboxylic Acid-13C,d3 Methyl Ester
CAS 1329634-37-8
Molekylær formel C1013CH7D3ClNO4
Mængde 10 mg
InChI formel InChI=1S/C11H10ClNO4/c1-13-8-4-6(12)3-7(11(15)16-2)10(8)17-5-9(13)14/h3-4H,5H2,1-2H3/i1+1D3
SMIL [2H][13C]([2H])([2H])N1C(=O)COc2c1cc(Cl)cc2C(=O)OC
IUPAC navn methyl 6-chloro-3-oxo-4-(trideuterio(1^{13}C)methyl)-1,4-benzoxazine-8-carboxylate
Molekylvægt (g/mol) 259.67
Formel vægt 259.052