missing translation for 'onlineSavingsMsg'
Få mere at vide

6-Chloro-3,4-dihydro-3-oxo-2H-1,4-benzoxazine-8-carboxylic Acid Methyl Ester, TRC

Artikelnummer. 30366602
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30366602 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30366602 Supplier Toronto Research Chemicals Supplier No. TRCC3652751G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 6-Chloro-3,4-dihydro-3-oxo-2H-1,4-benzoxazine-8-carboxylic Acid Methyl Ester
CAS 123040-75-5
Molekylær formel C10H8ClNO4
Mængde 1 g
InChI formel InChI=1S/C10H8ClNO4/c1-15-10(14)6-2-5(11)3-7-9(6)16-4-8(13)12-7/h2-3H,4H2,1H3,(H,12,13)
SMIL COC(=O)c1cc(Cl)cc2NC(=O)COc12
IUPAC navn methyl 6-chloro-3-oxo-4H-1,4-benzoxazine-8-carboxylate
Molekylvægt (g/mol) 241.63
Formel vægt 241.0142
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.