Få mere at vide
6-chloro-2-methyl-1,2,3,4-tetrahydroisoquinoline-1,3-dione, TRC
Artikelnummer. 30363248
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30363248 50 mg
30352616 5 mg
30349461 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30363248 Leverandør Toronto Research Chemicals Leverandørnr. TRCC37229550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 129689-05-0
Molekylær formel C10H8NO2Cl
Molekylvægt (g/mol) 209.63
IUPAC navn 6-chloro-2-methyl-4H-isoquinoline-1,3-dione
SMIL CN1C(=O)Cc2cc(Cl)ccc2C1=O
Tekniske data
Kemisk navn eller materiale 6-chloro-2-methyl-1,2,3,4-tetrahydroisoquinoline-1,3-dione
Mængde 50 mg
InChI formel InChI=1S/C10H8ClNO2/c1-12-9(13)5-6-4-7(11)2-3-8(6)10(12)14/h2-4H,5H2,1H3
Formel vægt 209.0244