missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2,4-dimethoxypyrimidine, 98+%, Thermo Scientific Chemicals

Catalog Number 11430287
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
6320-15-6
C6H7ClN2O2
174.58
MFCD00006068
JHNRTJRDRWKAIW-UHFFFAOYSA-N
6-chloro-2,4-dimethoxypyrimidine, 2,4-dimethoxy-6-chloropyrimidine, pyrimidine, 4-chloro-2,6-dimethoxy, 4-chloro-2,6-dimethoxy-pyrimidine, pubchem16350, acmc-209ndu, 4-chlor-2,6-dimethoxypyrimidin, 4-chloro-2,6-dimethoxypyrimidin, 2,6-dimethoxy-4-chloropyrimidine, 6-chloro-2,4-dimethixypyrimidine
80600
4-chloro-2,6-dimethoxypyrimidine
COC1=CC(Cl)=NC(OC)=N1
This item is not returnable. View return policy
6320-15-6
C6H7ClN2O2
174.58
MFCD00006068
JHNRTJRDRWKAIW-UHFFFAOYSA-N
6-chloro-2,4-dimethoxypyrimidine, 2,4-dimethoxy-6-chloropyrimidine, pyrimidine, 4-chloro-2,6-dimethoxy, 4-chloro-2,6-dimethoxy-pyrimidine, pubchem16350, acmc-209ndu, 4-chlor-2,6-dimethoxypyrimidin, 4-chloro-2,6-dimethoxypyrimidin, 2,6-dimethoxy-4-chloropyrimidine, 6-chloro-2,4-dimethixypyrimidine
80600
4-chloro-2,6-dimethoxypyrimidine
COC1=CC(Cl)=NC(OC)=N1

It is used in the synthesis of silyl and stannyl pyrimidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of silyl and stannyl pyrimidines.

Solubility
Soluble in chloroform, ethyl acetate and methanol.

Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point 71°C to 73°C
Quantity 1 g
Beilstein 137581
Sensitivity Hygroscopic
Solubility Information Soluble in chloroform,ethyl acetate and methanol.
Formula Weight 174.59
Percent Purity ≥98%
Chemical Name or Material 6-Chloro-2,4-dimethoxypyrimidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'