missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2,3-difluorobenzyl alcohol, 97%, Thermo Scientific™

Catalog Number p-7052475
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
887585-70-8
C7H5ClF2O
178.56
MFCD07368108
QOCZAGKTHBSPMS-UHFFFAOYSA-N
6-chloro-2,3-difluorobenzyl alcohol, 6-chloro-2,3-difluorobenzenemethanol, 6-chloro-2,3-difluorophenyl methanol
29922043
(6-chloro-2,3-difluorophenyl)methanol
OCC1=C(Cl)C=CC(F)=C1F
This item is not returnable. View return policy
887585-70-8
C7H5ClF2O
178.56
MFCD07368108
QOCZAGKTHBSPMS-UHFFFAOYSA-N
6-chloro-2,3-difluorobenzyl alcohol, 6-chloro-2,3-difluorobenzenemethanol, 6-chloro-2,3-difluorophenyl methanol
29922043
(6-chloro-2,3-difluorophenyl)methanol
OCC1=C(Cl)C=CC(F)=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 178.57
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Chloro-2,3-difluorobenzyl alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'