missing translation for 'onlineSavingsMsg'
Få mere at vide

6-Chloro[1,2,4]triazolo[4,3-b]pyridazin-3(2H)-one, TRC

Product Code. 30365510
Change view
Click to view available options
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30365510 500 mg
30368694 100 mg
30355416 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30365510 Supplier Toronto Research Chemicals Supplier No. TRCC364628500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33050-32-7
Molekylær formel C5H3N4OCl
Molekylvægt (g/mol) 170.56
IUPAC navn 6-chloro-2H-[1,2,4]triazolo[4,3-b]pyridazin-3-one
SMIL Oc1nnc2ccc(Cl)nn12

Specifications

Kemisk navn eller materiale 6-Chloro[1,2,4]triazolo[4,3-b]pyridazin-3(2H)-one
Mængde 500 mg
InChI formel InChI=1S/C5H3ClN4O/c6-3-1-2-4-7-8-5(11)10(4)9-3/h1-2H,(H,8,11)
Formel vægt 169.9995
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.