missing translation for 'onlineSavingsMsg'
Få mere at vide

MedChemExpress 6-CFDA

Artikelnummer. 16478896
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
100 mg
Pakningsstørrelse:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16478896 100 mg 100mg
16448758 10 mg -
16404677 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16478896 Leverandør MedChemExpress Leverandørnr. HYD0721

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress 6-CFDA is a fluorescent probe that can be used in flow cytometry and fluorescence microscopy. 6-CFDA also can be applied in intravital multiphoton microscopy to research hepatobiliary metabolism in animals.

Tekniske data

Indhold og opbevaring -20°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Produkttype Dye Reagents
Mængde 100 mg
Forsendelsestilstand Room temperature
Opløselighed DMSO : 100 mg/mL (217.21 mM; Need ultrasonic)
Mål HBV
Molekylvægt (g/mol) 460.39 g/mol
CAS 3348-03-6
Synonym 6-Carboxyfluorescein diacetate
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.