missing translation for 'onlineSavingsMsg'
Få mere at vide

6-Bromohexanal, TRC

Artikelnummer. 30328865
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30328865 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30328865 Leverandør Toronto Research Chemicals Leverandørnr. TRCB681878100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 6-Bromohexanal
CAS 57978-00-4
Molekylær formel C6H11OBr
Mængde 100 mg
InChI formel InChI=1S/C6H11BrO/c7-5-3-1-2-4-6-8/h6H,1-5H2
SMIL BrCCCCCC=O
IUPAC navn 6-bromohexanal
Molekylvægt (g/mol) 179.05
Formel vægt 177.9993
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.