missing translation for 'onlineSavingsMsg'
Learn More

6-Bromochromone-3-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7030990
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
51085-91-7
C10H5BrO4
269.05
MFCD01548933
IJTWNMVKNJSGMS-UHFFFAOYSA-N
6-bromochromone-3-carboxylic acid, 6-bromo-4-oxo-4h-chromene-3-carboxylic acid, 6-bromochromon-3-carboxylic acid
2756898
6-bromo-4-oxochromene-3-carboxylic acid
C1=CC2=C(C=C1Br)C(=O)C(=CO2)C(=O)O
This item is not returnable. View return policy
51085-91-7
C10H5BrO4
269.05
MFCD01548933
IJTWNMVKNJSGMS-UHFFFAOYSA-N
6-bromochromone-3-carboxylic acid, 6-bromo-4-oxo-4h-chromene-3-carboxylic acid, 6-bromochromon-3-carboxylic acid
2756898
6-bromo-4-oxochromene-3-carboxylic acid
C1=CC2=C(C=C1Br)C(=O)C(=CO2)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 191°C to 193°C
Quantity 1 g
Formula Weight 269.05
Percent Purity 97%
Chemical Name or Material 6-Bromochromone-3-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'