Få mere at vide
6-Bromobenzene-1,2,4-triol, TRC
Artikelnummer. 30335441
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30335441 100 mg
30334298 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30335441 Leverandør Toronto Research Chemicals Leverandørnr. TRCB795835100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Bromobenzene-1,2,4-triol
CAS 1480325-12-9
Molekylær formel C6H5BrO3
Mængde 100 mg
Synonym 6-Bromo-1,2,4-benzenetriol
InChI formel InChI=1S/C6H5BrO3/c7-4-1-3(8)2-5(9)6(4)10/h1-2,8-10H
SMIL Oc1cc(O)c(O)c(Br)c1
IUPAC navn 6-bromobenzene-1,2,4-triol
Molekylvægt (g/mol) 205.01
Formel vægt 203.9422