missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Bromo-8-iodoimidazo[1,2-a]pyridine, 95%, Alfa Aesar™
Brand: Alfa Aesar H37542.03
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
1364917-14-5 | |
322.931 | |
OKEMQQRZPHOOGS-UHFFFAOYSA-N | |
71299170 | |
C1=CN2C=C(C=C(C2=N1)I)Br |
C7H4BrIN2 | |
MFCD21609484 | |
6-bromo-8-iodoimidazo 1,2-a pyridine, 6-bromo-8-iodo-imidazo 1,2-a pyridine, 6-bromo-8-iodoimidazo 1,2-a pyridine,, imidazo 1,2-a pyridine,6-bromo-8-iodo, imidazo 1,2-a pyridine, 6-bromo-8-iodo | |
6-bromo-8-iodoimidazo[1,2-a]pyridine |
Specifications
6-Bromo-8-iodoimidazo[1,2-a]pyridine | |
Yellow | |
1g | |
C7H4BrIN2 | |
6-bromo-8-iodoimidazo 1,2-a pyridine, 6-bromo-8-iodo-imidazo 1,2-a pyridine, 6-bromo-8-iodoimidazo 1,2-a pyridine,, imidazo 1,2-a pyridine,6-bromo-8-iodo, imidazo 1,2-a pyridine, 6-bromo-8-iodo | |
C1=CN2C=C(C=C(C2=N1)I)Br | |
322.931 | |
322.92 | |
95% |
Light sensitive | |
147°C to 150°C | |
1364917-14-5 | |
MFCD21609484 | |
OKEMQQRZPHOOGS-UHFFFAOYSA-N | |
6-bromo-8-iodoimidazo[1,2-a]pyridine | |
71299170 | |
Powder |