missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one, 96%, Thermo Scientific Chemicals

Catalog Number p-7054110
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
355423-58-4
C8H5BrFNO2
246.04
MFCD20441932
AVHJMSMGASUDIT-UHFFFAOYSA-N
6-bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one, 6-bromo-7-fluoro-2h-benzo b 1,4 oxazin-3 4h-one, 2h-1,4-benzoxazin-3 4h-one, 6-bromo-7-fluoro, acmc-209ihm, 6-bromo-7-fluoro-4h-benzo 1,4 oxazin-3-one, 6-bromo-7-fluoro-2h-1,4-benzoxazin-3 4h-one, 6-bromo-7-fluoro-3,4-dihydro-2h-1,4-benzoxazin-3-one
23574320
6-bromo-7-fluoro-4H-1,4-benzoxazin-3-one
FC1=C(Br)C=C2NC(=O)COC2=C1
This item is not returnable. View return policy
355423-58-4
C8H5BrFNO2
246.04
MFCD20441932
AVHJMSMGASUDIT-UHFFFAOYSA-N
6-bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one, 6-bromo-7-fluoro-2h-benzo b 1,4 oxazin-3 4h-one, 2h-1,4-benzoxazin-3 4h-one, 6-bromo-7-fluoro, acmc-209ihm, 6-bromo-7-fluoro-4h-benzo 1,4 oxazin-3-one, 6-bromo-7-fluoro-2h-1,4-benzoxazin-3 4h-one, 6-bromo-7-fluoro-3,4-dihydro-2h-1,4-benzoxazin-3-one
23574320
6-bromo-7-fluoro-4H-1,4-benzoxazin-3-one
FC1=C(Br)C=C2NC(=O)COC2=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 246.03
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 6-Bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'