missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one, 96%

Brand:  Alfa Aesar™ H62026.MD

Additional Details : CAS Number : 355423-58-4 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15486936

  • 887.50 DKK / 250mg


Estimated Shipment
23-10-2020
Add to basket

Specifications

Chemical Identifiers

355423-58-4
246.035
AVHJMSMGASUDIT-UHFFFAOYSA-N
23574320
C1C(=O)NC2=CC(=C(C=C2O1)F)Br
C8H5BrFNO2
MFCD20441932
6-bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one, 6-bromo-7-fluoro-2h-benzo b 1,4 oxazin-3 4h-one, 2h-1,4-benzoxazin-3 4h-one, 6-bromo-7-fluoro, acmc-209ihm, 6-bromo-7-fluoro-4h-benzo 1,4 oxazin-3-one, 6-bromo-7-fluoro-2h-1,4-benzoxazin-3 4h-one, 6-bromo-7-fluoro-3,4-dihydro-2h-1,4-benzoxazin-3-one
6-bromo-7-fluoro-4H-1,4-benzoxazin-3-one

Specifications

6-Bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one
355423-58-4
MFCD20441932
AVHJMSMGASUDIT-UHFFFAOYSA-N
6-bromo-7-fluoro-4H-1,4-benzoxazin-3-one
23574320
Solid
250mg
C8H5BrFNO2
6-bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one, 6-bromo-7-fluoro-2h-benzo b 1,4 oxazin-3 4h-one, 2h-1,4-benzoxazin-3 4h-one, 6-bromo-7-fluoro, acmc-209ihm, 6-bromo-7-fluoro-4h-benzo 1,4 oxazin-3-one, 6-bromo-7-fluoro-2h-1,4-benzoxazin-3 4h-one, 6-bromo-7-fluoro-3,4-dihydro-2h-1,4-benzoxazin-3-one
C1C(=O)NC2=CC(=C(C=C2O1)F)Br
246.035
246.03
96%
Videos
SDS Documents