Få mere at vide
6-Bromo-6-dehydro-17Alpha-acetoxy Progesterone, TRC
Artikelnummer. 30136307
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136307 50 mg
30005687 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30136307 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68279550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 15251-04-4
Molekylær formel C23H29BrO4
Molekylvægt (g/mol) 449.38
Synonym Pregna-4,6-diene-3,20-dione, 17-(acetyloxy)-6-bromo-, Pregna-4,6-diene-3,20-dione, 6-bromo-17-hydroxy-, acetate (8CI), 17-(Acetyloxy)-6-bromopregna-4,6-diene-3,20-dione, 6-Bromo-6,7-didehydro-17α-acetoxyprogesterone
IUPAC navn [(8R,9S,10R,13S,14S,17R)-17-acetyl-6-bromo-10,13-dimethyl-3-oxo-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
SMIL CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=C(Br)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(=O)C
Tekniske data
Kemisk navn eller materiale 6-Bromo-6-dehydro-17a-acetoxy Progesterone
Mængde 50 mg
InChI formel InChI=1S/C23H29BrO4/c1-13(25)23(28-14(2)26)10-7-18-16-12-20(24)19-11-15(27)5-8-21(19,3)17(16)6-9-22(18,23)4/h11-12,16-18H,5-10H2,1-4H3/t16-,17+,18+,21-,22+,23+/m1/s1
Formel vægt 448.1249