Få mere at vide
6-Bromo-5-hydroxy-1-methyl-2-[(phenylthio)methyl]-1H-indole-3-carboxylic Acid Ethyl Ester, TRC
Artikelnummer. 30134993
Skift visning
Mængde:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134993 5 g
17937084 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134993 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6839055G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 131707-24-9
Molekylær formel C19 H18 Br N O3 S
Molekylvægt (g/mol) 420.32
Synonym 1-Methyl-2-phenylthiomethyl-3-ethoxycarbonyl-5-hydroxy-6-bromoindole, SI 5, SI 5
IUPAC navn ethyl 6-bromo-5-hydroxy-1-methyl-2-(phenylsulfanylmethyl)indole-3-carboxylate
SMIL O=C(C1=C(CSC2=CC=CC=C2)N(C)C3=C1C=C(C(Br)=C3)O)OCC
Tekniske data
Kemisk navn eller materiale 6-Bromo-5-hydroxy-1-methyl-2-[(phenylthio)methyl]-1H-indole-3-carboxylic Acid Ethyl Ester
Mængde 5 g
InChI formel InChI=1S/C19H18BrNO3S/c1-3-24-19(23)18-13-9-17(22)14(20)10-15(13)21(2)16(18)11-25-12-7-5-4-6-8-12/h4-10,22H,3,11H2,1-2H3
Formel vægt 419.0191