missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-3-pyridazinamine, 97%, Thermo Scientific™

Catalog Number p-3075832
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
88497-27-2
C4H4BrN3
174.001
MFCD01646092
FXTDHDQFLZNYKW-UHFFFAOYSA-N
2794779
6-bromopyridazin-3-amine
C1=CC(=NN=C1N)Br
This item is not returnable. View return policy
88497-27-2
C4H4BrN3
174.001
MFCD01646092
FXTDHDQFLZNYKW-UHFFFAOYSA-N
2794779
6-bromopyridazin-3-amine
C1=CC(=NN=C1N)Br
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 3-amino-6-bromopyridazine, 6-bromo-3-pyridazinamine, 6-bromo-pyridazin-3-ylamine, 2-amino-5-bromopyridazine, 3-pyridazinamine,6-bromo, 3-pyridazinamine, 6-bromo, 6-bromopyridazine-3-ylamine, 6-bromo-3-pyridazineamine, 3-bromo-6-aminopyridazine, acmc-20a0gf
Formula Weight 174
Percent Purity 97%
Chemical Name or Material 6-Bromo-3-pyridazinamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'