missing translation for 'onlineSavingsMsg'
Få mere at vide

6-Brom-3-hydroxypyrazin-2-carboxamid, TRC

Artikelnummer. 30138177
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138177 500 mg
30138072 250 mg
30007650 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30138177 Leverandør Toronto Research Chemicals Leverandørnr. TRCB103355500MG
 Begrænset vare
Forventet forsendelse: 20-08-2026
Læg i indkøbskurv
Add to basket
This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 259793-88-9
Molekylær formel C5 H4 Br N3 O2
Molekylvægt (g/mol) 218.008
Synonym 2-Pyrazinecarboxamide, 6-bromo-3,4-dihydro-3-oxo- (ACI), 6-Bromo-3,4-dihydro-3-oxo-2-pyrazinecarboxamide (ACI), Pyrazinecarboxamide, 6-bromo-3,4-dihydro-3-oxo- (9CI), 6-Bromo-3-hydroxypyrazine-2-carboxamide, Br-T-705
IUPAC navn 5-bromo-2-oxo-1H-pyrazin-3-carboxamid
SMIL NC(=O)C1=NC(=CNC1=O)Br

Specifications

Kemisk navn eller materiale 6-Bromo-3-hydroxypyrazine-2-carboxamide
Mængde 500 mg
InChI formel InChI=1S/C5H4BrN3O2/c6-2-1-8-5(11)3(9-2)4(7)10/h1H,(H2,7,10)(H,8,11)
Formel vægt 216.949
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.