Learn More
6-Bromo-1-hexene, 97%, ACROS Organics™
Brand: Acros Organics 364700050
Additional Details : CAS Number : 2695-47-8 Weight : 0.05500kg Transport : UN number : 1993 Chem class : 3 Pack group : III
Packaging | Glass bottle |
---|---|
Quantity | 5g |
Chemical Identifiers
2695-47-8 | |
163.06 | |
RIMXEJYJXDBLIE-UHFFFAOYSA-N | |
75906 | |
C=CCCCCBr |
C6H11Br | |
MFCD00000269 | |
6-bromo-1-hexene, 5-hexenyl bromide, 1-hexene, 6-bromo, 1-bromo-5-hexene, 6-bromohexene, 6-bromo-hexene, 6-bromo-hex-1-ene, l-bromo-5-hexene, 6-bromo-1-hexen, 1-bromohex-5-ene | |
6-bromohex-1-ene |
Specifications
6-Bromo-1-hexene | |
52°C | |
Glass bottle | |
1.217 | |
Yellow | |
2695-47-8 | |
100.0 | |
C6H11Br | |
MFCD00000269 | |
RIMXEJYJXDBLIE-UHFFFAOYSA-N | |
6-bromohex-1-ene | |
75906 | |
Liquid |
1.2170g/mL | |
Authentic | |
1.4645 to 1.4665 (20°C, 589nm) | |
47.0°C to 51.0°C (16.0 mmHg) | |
5g | |
96.0 | |
96% min. (GC) | |
Br(CH2)4CH=CH2 | |
6-bromo-1-hexene, 5-hexenyl bromide, 1-hexene, 6-bromo, 1-bromo-5-hexene, 6-bromohexene, 6-bromo-hexene, 6-bromo-hex-1-ene, l-bromo-5-hexene, 6-bromo-1-hexen, 1-bromohex-5-ene | |
C=CCCCCBr | |
163.06 | |
163.06 | |
97% |
Safety and Handling
- 6-Bromo-1-hexene
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.