Learn More
6-Bromo-1,4-benzodioxane, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
6-Bromo-1,4-benzodioxane is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Bromo-1,4-benzodioxane is used as a pharmaceutical intermediate.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
6-Bromo-1,4-benzodioxane is used as a pharmaceutical intermediate.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
Density | 1.598 |
Boiling Point | 259°C to 260°C |
Flash Point | >110°C (230°F) |
Refractive Index | 1.589 |
Quantity | 1 g |
Beilstein | 6223 |
Formula Weight | 215.05 |
Percent Purity | 98% |
Chemical Name or Material | 6-Bromo-1,4-benzodioxane |