Få mere at vide
Tocris Bioscience™ 6-Bnz-cAMP sodium salt
GREENER_CHOICE
Cell permeable cAMP analog
Brand:  Tocris Bioscience™ 5255/1
Encompass
Produktkode 15726626
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
6-Bnz-cAMP sodium salt is a cell permeable cAMP analog; selectively activates cAMP-dependent PKA but not Epac signaling pathways. Acts synergistically with 8-CPT-2Me-cAMP (Cat. No. 1645) to inhibit vascular smooth muscle cell proliferation.
Tekniske data
Tekniske data
98%
1135306-29-4
1 mg
N-Benzoyladenosine cyclic 3 ',5 '-(hydrogen phosphate) monosodium salt
-20°C
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Produkttitel
Vælg et problem
For Research Use Only