missing translation for 'onlineSavingsMsg'
Learn More

6-Benzyloxypyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific™

Catalog Number p-7030161
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
832735-54-3
C18H22BNO3
311.188
MFCD06798263
MXNLRVZITPPZHT-UHFFFAOYSA-N
2-benzyloxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-benzyloxypyridine-5-boronic acid, pinacol ester, 6-benzyloxy pyridine-3-boronic acid pinacol ester, 2-benzyloxypyridine-5-boronic acid pinacol ester, 2-benzyloxy-5-pyridineboronic acid pinacol ester, 2-benzyloxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, amtb159, 2-benzyloxypyridine-5-boronic acid,pinacol ester, 6-benzyloxy pyridine-3-boronicacidpinacolester, 2-benzyloxy-pyridine-5-boronic acid pinacol ester
17750217
2-phenylmethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)OCC3=CC=CC=C3
This item is not returnable. View return policy
832735-54-3
C18H22BNO3
311.188
MFCD06798263
MXNLRVZITPPZHT-UHFFFAOYSA-N
2-benzyloxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-benzyloxypyridine-5-boronic acid, pinacol ester, 6-benzyloxy pyridine-3-boronic acid pinacol ester, 2-benzyloxypyridine-5-boronic acid pinacol ester, 2-benzyloxy-5-pyridineboronic acid pinacol ester, 2-benzyloxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, amtb159, 2-benzyloxypyridine-5-boronic acid,pinacol ester, 6-benzyloxy pyridine-3-boronicacidpinacolester, 2-benzyloxy-pyridine-5-boronic acid pinacol ester
17750217
2-phenylmethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)OCC3=CC=CC=C3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 138°C to 142°C
Quantity 250 mg
Formula Weight 311.19
Percent Purity 95%
Chemical Name or Material 6-Benzyloxypyridine-3-boronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'