missing translation for 'onlineSavingsMsg'
Få mere at vide

6-Azido-1,6-dideoxy-3,4-O-isopropylidene-D-lyxo-2-hexulofuranose, TRC

Artikelnummer. 30335457
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30335457 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335457 Supplier Toronto Research Chemicals Supplier No. TRCA88456025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 6-Azido-1,6-dideoxy-3,4-O-isopropylidene-D-lyxo-2-hexulofuranose
CAS 122194-05-2
Molekylær formel C9H15N3O4
Mængde 25 mg
Synonym 6-Azido-1,6-dideoxy-3,4-O-(1-methylethylidene)-D-tagatose
InChI formel InChI=1S/C9H15N3O4/c1-8(2)15-6-5(4-11-12-10)14-9(3,13)7(6)16-8/h5-7,13H,4H2,1-3H3/t5-,6+,7+,9?/m1/s1
SMIL CC1(C)O[C@H]2[C@@H](CN=[N+]=[N-])OC(C)(O)[C@H]2O1
IUPAC navn (3aS,6R,6aS)-6-(azidomethyl)-2,2,4-trimethyl-6,6a-dihydro-3aH-furo[3,4-d][1,3]dioxol-4-ol
Molekylvægt (g/mol) 229.23
Formel vægt 229.1063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.