missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Azauracil, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
6-Azauracil (6-AU) is used as a growth inhibitor of various microorganisms via depletion of intracellular GTP and UTP nucleotide pools. 6-Azauracil has been widely used in investigations on modulation of transcription, especially in yeast models.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Azauracil (6-AU) is used as a growth inhibitor of various microorganisms via depletion of intracellular GTP and UTP nucleotide pools. 6-Azauracil has been widely used in investigations on modulation of transcription, especially in yeast models.
Solubility
Partly soluble in water, ethanol, DMSO, and 1 M NH4OH (50 mg/ml).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
6-Azauracil (6-AU) is used as a growth inhibitor of various microorganisms via depletion of intracellular GTP and UTP nucleotide pools. 6-Azauracil has been widely used in investigations on modulation of transcription, especially in yeast models.
Solubility
Partly soluble in water, ethanol, DMSO, and 1 M NH4OH (50 mg/ml).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 274°C to 282°C |
Quantity | 5 g |
Beilstein | 116472 |
Solubility Information | Partly soluble in water,ethanol,DMSO,and 1 M NH4OH (50mg/ml). |
Formula Weight | 113.08 |
Percent Purity | 98% |
Chemical Name or Material | 6-Azauracil |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'