Få mere at vide
6-Amino-5-nitroso-3-methyluracil, TRC
Artikelnummer. 30134667
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134667 100 mg
30008377 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134667 Leverandør Toronto Research Chemicals Leverandørnr. TRCA618445100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 61033-04-3
Molekylær formel C5 H6 N4 O3
Molekylvægt (g/mol) 170.13
Synonym 2,4(1H,3H)-Pyrimidinedione, 6-amino-3-methyl-5-nitroso- (9CI, ACI), 6-Amino-3-methyl-5-nitroso-2,4(1H,3H)-pyrimidinedione (ACI), Uracil, 6-amino-3-methyl-5-nitroso- (6CI), 6-Amino-3-methyl-5-nitrosopyrimidine-2,4-dione, 6-Amino-5-nitroso-3-methyluracil
IUPAC navn 6-amino-3-methyl-5-nitroso-1H-pyrimidine-2,4-dione
SMIL CN1C(=O)NC(=C(N=O)C1=O)N
Tekniske data
Kemisk navn eller materiale 6-Amino-5-nitroso-3-methyluracil
Analyt- eller komponentnavne 6-Amino-5-nitroso-3-methyluracil
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C5H6N4O3/c1-9-4(10)2(8-12)3(6)7-5(9)11/h6H2,1H3,(H,7,11)
Løsningstype Neat
Industry Type Forensics
Mængde 100 mg
Formel vægt 170.044