Få mere at vide
6-Amino-5-nitroso-3-methyluracil-d3, TRC
Artikelnummer. 30326564
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30326564 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326564 Leverandør Toronto Research Chemicals Leverandørnr. TRCA61844750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Amino-5-nitroso-3-methyluracil-d3
CAS 1246819-56-6
Molekylær formel C5H3D3N4O3
Mængde 50 mg
Synonym 6-Amino-3-methyl-5-nitrosopyrimidine-2,4-dione-d3
InChI formel InChI=1S/C5H6N4O3/c1-9-4(10)2(8-12)3(6)7-5(9)11/h6H2,1H3,(H,7,11)/i1D3
SMIL [2H]C([2H])([2H])N1C(=O)NC(=C(N=O)C1=O)N
IUPAC navn 6-amino-5-nitroso-3-(trideuteriomethyl)-1H-pyrimidine-2,4-dione
Molekylvægt (g/mol) 173.14
Formel vægt 173.0628