Få mere at vide
6-Amino-3-methyluracil-d3, TRC
Artikelnummer. 30324157
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30324157 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324157 Leverandør Toronto Research Chemicals Leverandørnr. TRCA61827710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Amino-3-methyluracil-d3
CAS 1246820-43-8
Molekylær formel C5H4D3N3O2
Mængde 10 mg
Synonym 6-Amino-3-methylpyrimidine-2,4-(1H,3H)-dione-d3, 3-Methyl-6-aminouracil-d3
InChI formel InChI=1S/C5H7N3O2/c1-8-4(9)2-3(6)7-5(8)10/h2H,6H2,1H3,(H,7,10)/i1D3
SMIL [2H]C([2H])([2H])N1C(=O)NC(=CC1=O)N
IUPAC navn 6-amino-3-(trideuteriomethyl)-1H-pyrimidine-2,4-dione
Molekylvægt (g/mol) 144.15
Formel vægt 144.0727