Learn More
6,9-Dichloro-2-methoxyacridine, 98%, Thermo Scientific Chemicals
Catalog Number p-7044333
Click to view available options
Quantity:
5 g
25 g
86-38-4
C14H9Cl2NO
278.132
MFCD00005028
RYRNQWYNHLLOGX-UHFFFAOYSA-N
halocrin, 3,9-dichloro-7-methoxyacridine, halocrine, 6.9-dichloro-2-methoxyacridine, 2-methoxy-6,9-dichloroacridine, acridine,9-dichloro-2-methoxy, acridine, 6,9-dichloro-2-methoxy, bidd:gt0206, 6,9dichloro-2-methoxyacridine, 6,9-dichloro-2-methoxy-acridine
66577
6,9-dichloro-2-methoxyacridine
COC1=CC2=C(C3=C(C=C(C=C3)Cl)N=C2C=C1)Cl
86-38-4
C14H9Cl2NO
278.132
MFCD00005028
RYRNQWYNHLLOGX-UHFFFAOYSA-N
halocrin, 3,9-dichloro-7-methoxyacridine, halocrine, 6.9-dichloro-2-methoxyacridine, 2-methoxy-6,9-dichloroacridine, acridine,9-dichloro-2-methoxy, acridine, 6,9-dichloro-2-methoxy, bidd:gt0206, 6,9dichloro-2-methoxyacridine, 6,9-dichloro-2-methoxy-acridine
66577
6,9-dichloro-2-methoxyacridine
COC1=CC2=C(C3=C(C=C(C=C3)Cl)N=C2C=C1)Cl
6,9-Dichloro-2-methoxyacridine is used to prepare 6-chloro-2-methoxy-acridin-9-ylamine by reacting with phenol and ammonium carbonate. It is used in the preparation of 9-amino-6-chloro-2-methoxyacridine, N'-(6-Chloro-2-methoxy-acridin-9-yl)-heptylamine and N,N'-bis-(6-chloro-2-methoxy-acridin-9-yl)-hexane-1,6-diamine. Further, it is involved in the preparation of 4-aminoquinoline and 9-aminoacridine derivatives by reaction with quinolizidinylalkylamines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6,9-Dichloro-2-methoxyacridine is used to prepare 6-chloro-2-methoxy-acridin-9-ylamine by reacting with phenol and ammonium carbonate. It is used in the preparation of 9-amino-6-chloro-2-methoxyacridine, N′-(6-Chloro-2-methoxy-acridin-9-yl)-heptylamine and N,N′-bis-(6-chloro-2-methoxy-acridin-9-yl)-hexane-1,6-diamine. Further, it is involved in the preparation of 4-aminoquinoline and 9-aminoacridine derivatives by reaction with quinolizidinylalkylamines.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 163°C to 165°C
Quantity 5 g
Beilstein 226309
Solubility Information Insoluble in water.
Formula Weight 278.14
Percent Purity 98%
Chemical Name or Material 6,9-Dichloro-2-methoxyacridine