Få mere at vide
6,8-Difluoro-7-hydroxy-4-methylcoumarin (DIFMU), TRC
Artikelnummer. 30136894
Skift visning
Mængde:
2.5 mg
5 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30136894 5 mg -
30132643 25 mg -
17959444 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136894 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4524485MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 215868-23-8
Molekylær formel C10H6F2O3
Molekylvægt (g/mol) 212.15
SMIL CC1=CC(=O)Oc2c(F)c(O)c(F)cc12
Tekniske data
Kemisk navn eller materiale 6,8-Difluoro-7-hydroxy-4-methylcoumarin (DIFMU)