Få mere at vide
6,8-Dibromo-3-cyclohexyl-3,4-dihydro-2(1H)-quinazolinone, Mikromol™
Artikelnummer. 30052459
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052459 25 mg
30049667 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052459 Leverandør Mikromol Leverandørnr. MM0065.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6,8-Dibromo-3-cyclohexyl-3,4-dihydro-2(1H)-quinazolinone
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6,8-Dibromo-3-cyclohexyl-3,4-dihydro-2(1H)-quinazolinone
CAS 21237-84-3
Analyt- eller komponentnavne 6,8-Dibromo-3-cyclohexyl-3,4-dihydro-2(1H)-quinazolinone
Forsendelsestilstand Room Temperature
Molekylær formel C14 H16 Br2 N2 O
Certificeringer/compliance ISO 9001
Synonym 6,8-Dibromo-3-cyclohexyl-3,4-dihydro-2(1H)-quinazolinone
InChI formel InChI=1S/C14H16Br2N2O/c15-10-6-9-8-18(11-4-2-1-3-5-11)14(19)17-13(9)12(16)7-10/h6-7,11H,1-5,8H2,(H,17,19)
SMIL Brc1cc(Br)c2NC(=O)N(Cc2c1)C3CCCCC3
IUPAC navn 6,8-dibromo-3-cyclohexyl-1,4-dihydroquinazolin-2-one
Molekylvægt (g/mol) 388.1
Mængde 25 mg
Formel vægt 385.9629 g/mol