missing translation for 'onlineSavingsMsg'
Få mere at vide

6,7-Dimethoxy-3,4-dihydronaphthalen-1(2H)-one, 97%, BLD Pharm™

Artikelnummer. 31087673
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
250 mg
1 g
5 g
10 g
25 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
31087673 250 mg
31076540 25 g
31078733 100 mg
31081185 10 g
31081462 1 g
31087150 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31087673 Supplier BLD Pharm Supplier No. BD14011250mg
 Begrænset vare
Forventet forsendelse: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Kemisk navn eller materiale 6,7-Dimethoxy-3,4-dihydronaphthalen-1(2H)-one
CAS 13575-75-2
Molekylær formel C12H14O3
MDL nummer MFCD00134100
Mængde 250 mg
SMIL O=C1CCCC2=C1C=C(OC)C(OC)=C2
Molekylvægt (g/mol) 206.24
Procent renhed 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.