Få mere at vide
6,7-Dihydroxyflavone, TRC
Artikelnummer. 30179858
Skift visning
Mængde:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179858 5 mg
30180054 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30179858 Leverandør Toronto Research Chemicals Leverandørnr. TRCD6790085MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 38183-04-9
Molekylær formel C15 H10 O4
Molekylvægt (g/mol) 254.24
IUPAC navn 6,7-dihydroxy-2-phenyl-chromen-4-one
SMIL Oc1cc2OC(=CC(=O)c2cc1O)c3ccccc3
Tekniske data
Kemisk navn eller materiale 6,7-Dihydroxyflavone
Mængde 5 mg
InChI formel InChI=1S/C15H10O4/c16-11-7-14(9-4-2-1-3-5-9)19-15-8-13(18)12(17)6-10(11)15/h1-8,17-18H
Formel vægt 254.0579