Få mere at vide
6,7-Dihydrothieno[3,2-c]-pyridin-4(5H)-one, Mikromol™
Artikelnummer. 30049606
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049606 25 mg
30049072 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049606 Leverandør Mikromol Leverandørnr. MM0150.060025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6,7-Dihydrothieno[3,2-c]-pyridin-4(5H)-one
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6,7-Dihydrothieno[3,2-c]-pyridin-4(5H)-one
CAS 68559-60-4
Analyt- eller komponentnavne 6,7-Dihydrothieno[3,2-c]-pyridin-4(5H)-one
Forsendelsestilstand Room Temperature
Molekylær formel C7 H7 N O S
Certificeringer/compliance ISO 17025
Synonym Ticlopidine Hydrochloride Imp. B (EP), Ticlopidine Imp. B (EP), Ticlopidine USP Related Compound A, Ticlopidine USP RC A, 6,7-Dihydrothieno[3,2-c]-pyridin-4(5H)-one, Ticlopidine Related Compound A, Ticlopidine Hydrochloride Impurity B, Ticlopidine Impurity B
InChI formel InChI=1S/C7H7NOS/c9-7-5-2-4-10-6(5)1-3-8-7/h2,4H,1,3H2,(H,8,9)
SMIL O=C1NCCc2sccc12
IUPAC navn 6,7-dihydro-5H-thieno[3,2-c]pyridin-4-one
Molekylvægt (g/mol) 153.2
Mængde 25 mg
Formel vægt 153.0248 g/mol