Få mere at vide
6,7-Dihydro-2-(1-pyrrolidinyl)-5H-pyrrolo[3,4-d]pyrimidine Dihydrochloride;, TRC
Artikelnummer. 30418614
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30418614 100 mg
30410143 1 g
30413587 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30418614 Leverandør Toronto Research Chemicals Leverandørnr. TRCP184751100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1864058-89-8
Molekylær formel C10H14N4 . 2(HCl)
Molekylvægt (g/mol) 263.16
Synonym 1-[5H,6H,7H-Pyrrolo[3,4-d]pyrimidin-2-yl]pyrrolidine Dihydrochloride
IUPAC navn 2-pyrrolidin-1-yl-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidine;dihydrochloride
SMIL Cl.Cl.C1CCN(C1)c2ncc3CNCc3n2
Tekniske data
Kemisk navn eller materiale 6,7-Dihydro-2-(1-pyrrolidinyl)-5H-pyrrolo[3,4-d]pyrimidine Dihydrochloride
Mængde 100 mg
InChI formel InChI=1S/C10H14N4.2ClH/c1-2-4-14(3-1)10-12-6-8-5-11-7-9(8)13-10;;/h6,11H,1-5,7H2;2*1H
Formel vægt 262.0752