missing translation for 'onlineSavingsMsg'
Få mere at vide

6,7-Didehydronorethisterone Acetate, Mikromol™

Artikelnummer. 30459105
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30459105 4 x 25 mg
30050686 25 mg
30049649 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 30459105 Leverandør Mikromol Leverandørnr. MM0219.0400254AMP

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

6,7-Didehydronorethisterone Acetate, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 6,7-Didehydronorethisterone Acetate
CAS 106765-04-2
Analyt- eller komponentnavne 6,7-Didehydronorethisterone Acetate
Forsendelsestilstand Room Temperature
Molekylær formel C22 H26 O3
Certificeringer/compliance ISO 17025
Synonym 6,7-Didehydronorethisterone Acetate
InChI formel InChI=1S/C22H26O3/c1-4-22(25-14(2)23)12-10-20-19-7-5-15-13-16(24)6-8-17(15)18(19)9-11-21(20,22)3/h1,5,7,13,17-20H,6,8-12H2,2-3H3/t17-,18+,19+,20-,21-,22-/m0/s1
Løsningstype Neat
SMIL CC(=O)O[C@]1(CC[C@H]2[C@@H]3C=CC4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C)C#C
IUPAC navn [(8R,9S,10R,13S,14S,17R)-17-ethynyl-13-methyl-3-oxo-1,2,8,9,10,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl] acetate
Molekylvægt (g/mol) 338.44
Mængde 4 x 25 mg
Formel vægt 338.1882 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.