Få mere at vide
6,7-Demethylene-6,7-dehydro Drospirenone, TRC
Artikelnummer. 30134279
Skift visning
Mængde:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134279 10 mg
30134132 100 mg
30012114 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134279 Leverandør Toronto Research Chemicals Leverandørnr. TRCD23103010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 67372-69-4
Molekylær formel C23 H28 O3
Molekylvægt (g/mol) 352.47
Synonym (15α,16α,17α)-15,16-dihydro-17-hydroxy-3-oxo-3'H-Cyclopropa[15,16]pregna-4,6,15-triene-21-carboxylic acid γ-lactone, 17β-Hydroxy-15β,16β-methylene-3-oxo-17α-pregna-4,6-diene-21-carboxylic acid γ-lactone, Drospirenone Imp. D (EP), 3'H-Cyclopropa[15,16]pregna-4,6,15-triene-21-carboxylic acid, 15,16-dihydro-17-hydroxy-3-oxo-, γ-lactone, (15α,16α,17α)-
SMIL C[C@]12CCC(=O)C=C1C=C[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3[C@@H]5C[C@@H]5[C@@]46CCC(=O)O6
Tekniske data
Kemisk navn eller materiale delta6-Drospirenone
Mængde 10 mg
InChI formel InChI=1S/C23H28O3/c1-21-8-5-14(24)11-13(21)3-4-15-17(21)6-9-22(2)20(15)16-12-18(16)23(22)10-7-19(25)26-23/h3-4,11,15-18,20H,5-10,12H2,1-2H3/t15-,16-,17+,18+,20-,21+,22+,23+/m1/s1
Formel vægt 352.2038