Få mere at vide
6,6'-Oxybis-2-naphthalenesulfonic Acid Disodium Salt >90%, TRC
Artikelnummer. 17941745
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17941745 100 mg
17940615 50 mg
17932405 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17941745 Leverandør Toronto Research Chemicals Leverandørnr. TRCO867500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 61551-82-4
Molekylær formel C20 H12 O7 S2 . 2 Na
Molekylvægt (g/mol) 474.41
Synonym 6,6'-oxybis-2-naphthalenesulfonic acid disodium salt (9CI), Disodium 6,6'-oxybis(2-naphthalenesulfonate)
IUPAC navn disodium;6-(6-sulfonatonaphthalen-2-yl)oxynaphthalene-2-sulfonate
SMIL [Na+].[Na+].[O-]S(=O)(=O)c1ccc2cc(Oc3ccc4cc(ccc4c3)S(=O)(=O)[O-])ccc2c1
Tekniske data
Kemisk navn eller materiale Disodium 6,6'-Oxybis(2-naphthalenesulfonate)
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C20H14O7S2.2Na/c21-28(22,23)19-7-3-13-9-17(5-1-15(13)11-19)27-18-6-2-16-12-20(29(24,25)26)8-4-14(16)10-18;;/h1-12H,(H,21,22,23)(H,24,25,26);;/q;2*+1/p-2
Formel vægt 473.982
Procent renhed >95%