Få mere at vide
6,6-Dimethyl-2-propyl-3-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-3,6-dihydro-4H-furo[3,4-d]imidazol-4-one, Mikromol™
Artikelnummer. 30458679
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30458679 4 x 25 mg
30040551 25 mg
17958934 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30458679 Leverandør Mikromol Leverandørnr. MM1317.0200254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6,6-Dimethyl-2-propyl-3-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-3,6-dihydro-4H-furo[3,4-d]imidazol-4-one, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-{[2'-(1H-Tetrazol-5-yl)biphenyl-4-yl]methyl}-4,4-dimethyl-2-propyl-1H-furo[3,4-d]imidazol-6(4H)-one
CAS 849206-43-5
Analyt- eller komponentnavne 1-{[2'-(1H-Tetrazol-5-yl)biphenyl-4-yl]methyl}-4,4-dimethyl-2-propyl-1H-furo[3,4-d]imidazol-6(4H)-one
Forsendelsestilstand Room Temperature
Molekylær formel C24 H24 N6 O2
Certificeringer/compliance ISO 17025
Synonym 6,6-Dimethyl-2-propyl-3-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-3,6-dihydro-4H-furo[3,4-d]imidazol-4-one, Olmesartan Medoxomil Imp. B (EP), 4H-Furo[3,4-d]imidazol-4-one, 3,6-dihydro-6,6-dimethyl-2-propyl-3-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, 4H-Furo[3,4-d]imidazol-4-one, 3,6-dihydro-6,6-dimethyl-2-propyl-3-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]- (9CI), 1-{[2'-(1H-Tetrazol-5-yl)biphenyl-4-yl]methyl}-4,4-dimethyl-2-propyl-1H-furo[3,4-d]imidazol-6(4H)-one, Olmesartan Medoxomil RC A (USP), Olmesartan Medoxomil Related Compound A, Olmesartan Medoxomil Impurity B
InChI formel InChI=1S/C24H24N6O2/c1-4-7-19-25-21-20(23(31)32-24(21,2)3)30(19)14-15-10-12-16(13-11-15)17-8-5-6-9-18(17)22-26-28-29-27-22/h5-6,8-13H,4,7,14H2,1-3H3,(H,26,27,28,29)
Løsningstype Neat
SMIL CCCc1nc2c(C(=O)OC2(C)C)n1Cc3ccc(cc3)c4ccccc4c5nnn[nH]5
IUPAC navn 6,6-dimethyl-2-propyl-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]furo[3,4-d]imidazol-4-one
Molekylvægt (g/mol) 428.49
Mængde 4 x 25 mg
Formel vægt 428.1961 g/mol