missing translation for 'onlineSavingsMsg'
Få mere at vide

6,6’-Dihydroxy-5,5’-dimethoxy-2,2’-diphenic Acid Dimethyl Ester, TRC

Artikelnummer. 30386793
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30386793 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30386793 Leverandør Toronto Research Chemicals Leverandørnr. TRCD45270510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 6,6'-Dihydroxy-5,5'-dimethoxy-2,2'-diphenic Acid Dimethyl Ester
CAS 1173188-30-1
Molekylær formel C18H18O8
Mængde 10 mg
Synonym 6,6'-Dihydroxy-5,5'-dimethoxy-[1,1'-biphenyl]-2,2'-dicarboxylic Acid 2,2'-Dimethyl Ester
InChI formel InChI=1S/C18H18O8/c1-23-11-7-5-9(17(21)25-3)13(15(11)19)14-10(18(22)26-4)6-8-12(24-2)16(14)20/h5-8,19-20H,1-4H3
SMIL COC(=O)c1ccc(OC)c(O)c1c2c(O)c(OC)ccc2C(=O)OC
IUPAC navn methyl 3-hydroxy-2-(2-hydroxy-3-methoxy-6-methoxycarbonylphenyl)-4-methoxybenzoate
Molekylvægt (g/mol) 362.33
Formel vægt 362.1002
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.