missing translation for 'onlineSavingsMsg'
Learn More

6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)chroman, 97%, Thermo Scientific™

Catalog Number p-3882038
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
1002727-88-9
C15H21BO3
260.14
MFCD12028565
NDSHAELUPJMEBM-UHFFFAOYSA-N
2-chroman-6-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl chroman, 2-3,4-dihydro-2h-1-benzopyran-6-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, chroman-6-boronic acid, pinacol ester, amtb667, chroman-6-boronic acid pinacol ester, chroman-6-ylboronic acid pinacol ester, 3,4-dihydro-2h-chromene-6-boronic acid, pinacol ester, 3,4-dihydro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-chromene
43811034
2-(3,4-dihydro-2H-chromen-6-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=CC2=C(OCCC2)C=C1
This item is not returnable. View return policy
1002727-88-9
C15H21BO3
260.14
MFCD12028565
NDSHAELUPJMEBM-UHFFFAOYSA-N
2-chroman-6-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl chroman, 2-3,4-dihydro-2h-1-benzopyran-6-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, chroman-6-boronic acid, pinacol ester, amtb667, chroman-6-boronic acid pinacol ester, chroman-6-ylboronic acid pinacol ester, 3,4-dihydro-2h-chromene-6-boronic acid, pinacol ester, 3,4-dihydro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-chromene
43811034
2-(3,4-dihydro-2H-chromen-6-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=CC2=C(OCCC2)C=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 260.14
Percent Purity 97%
Chemical Name or Material 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)chroman
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'