Få mere at vide
6-[4-(1-Cyclohexyl-1H-tetrazol-5-yl)butoxy]-1-[4-(1-cyclohexyl-1H-tetrazol-5-yl)butyl]-3,4-dihydro-2(1H)-quinolinone, TRC
Artikelnummer. 30180316
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180316 50 mg
30179902 10 mg
30180027 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30180316 Leverandør Toronto Research Chemicals Leverandørnr. TRCC98825550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 865792-18-3
Molekylær formel C31 H45 N9 O2
Molekylvægt (g/mol) 575.75
Synonym 2(1H)-Quinolinone, 6-[4-(1-cyclohexyl-1H-tetrazol-5-yl)butoxy]-1-[4-(1-cyclohexyl-1H-tetrazol-5-yl)butyl]-3,4-dihydro-, 6-[4-(1-Cyclohexyl-1H-tetrazol-5-yl)butoxy]-1-[4-(1-cyclohexyl-1H-tetrazol-5-yl)butyl]-3,4-dihydro-2(1H)-quinolinone, 1-(4-(5-Cyclohexyl-1H-tetrazol-1-yl)butyl)-6-(4-(1-cyclohexyl-1H-tetrazol-5-yl)butoxy)-3,4-dihydroquinolin-2(1H)-one
IUPAC navn 6-[4-(1-cyclohexyltetrazol-5-yl)butoxy]-1-[4-(1-cyclohexyltetrazol-5-yl)butyl]-3,4-dihydroquinolin-2-one
SMIL O=C1CCc2cc(OCCCCc3nnnn3C4CCCCC4)ccc2N1CCCCc5nnnn5C6CCCCC6
Tekniske data
Kemisk navn eller materiale 1-(4-(5-Cyclohexyl-1H-tetrazol-1-yl)butyl)-6-(4-(1-cyclohexyl-1H-tetrazol-5-yl)butoxy)-3,4-dihydroquinolin-2(1H)-one
Mængde 50 mg
InChI formel InChI=1S/C31H45N9O2/c41-31-20-17-24-23-27(42-22-10-8-16-30-33-35-37-40(30)26-13-5-2-6-14-26)18-19-28(24)38(31)21-9-7-15-29-32-34-36-39(29)25-11-3-1-4-12-25/h18-19,23,25-26H,1-17,20-22H2
Formel vægt 575.3696