Få mere at vide
6-(3-Pyridinylcarbonyl)valerolactam-d4, TRC
Artikelnummer. 30419283
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30419283 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30419283 Leverandør Toronto Research Chemicals Leverandørnr. TRCP99167710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-(3-Pyridinylcarbonyl)valerolactam-d4
CAS 1189467-35-3
Molekylær formel C11H8D4N2O2
Mængde 10 mg
Synonym 3-[3-(Pyridinyl-d4)carbonyl]-2-piperidinone
InChI formel InChI=1S/C11H12N2O2/c14-10(8-3-1-5-12-7-8)9-4-2-6-13-11(9)15/h1,3,5,7,9H,2,4,6H2,(H,13,15)/i1D,3D,5D,7D
SMIL [2H]c1nc([2H])c(C(=O)C2CCCNC2=O)c([2H])c1[2H]
IUPAC navn 3-(2,4,5,6-tetradeuteriopyridine-3-carbonyl)piperidin-2-one
Molekylvægt (g/mol) 208.25
Formel vægt 208.115